Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Human Recombinant Histone H2B

Catalog No. 03185MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-185MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-185MI Supplier MilliporeSigma™ Supplier No. 03185
Add to Cart
Add to Cart

Specifications

Characterization >95% by SDS-PAGE
For Use With (Application) Western Blot, Kinase Assay, DNA-binding Proteins, Chromatin Assembly Studies
Formulation Liquid in Dulbecco's PBS
Name Histone H2B
Purification Method SDS-PAGE
Quantity 50 μg
Source E. coli
Storage Requirements -80°C for 12 months
Cross Reactivity Human
Recombinant Recombinant
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.