Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Human Recombinant LSD1

Catalog No. 03229MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-229MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-229MI Supplier MilliporeSigma™ Supplier No. 03229
Add to Cart
Add to Cart

Specifications

Characterization >90% by SDS-PAGE
For Use With (Application) Enzyme Assay (Demethylation enzyme assays)
Formulation 6mM Na2HPO4, 1.1mM K2HPO4, pH 7.2, 82mM sodium chloride, 1.6mM KCl and 40% glycerol.
Name LSD1
Quantity 50 μg
Source E. coli
Storage Requirements -80°C for 12 months
Cross Reactivity Human
Recombinant Recombinant
Form Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.