Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Human Recombinant p300 (Catalytic Domain)

Catalog No. 03188MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-188MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-188MI Supplier MilliporeSigma™ Supplier No. 03188
Add to Cart
Add to Cart

Specifications

Characterization >75% by SDS-PAGE
For Use With (Application) Enzyme Assay (Acetyltransferase assay)
Formulation In liquid containing 50mM Tris/HCl, pH 8.0, 0.1mM EDTA, 10% glycerol
Name p300 (Catalytic Domain)
Purification Method SDS-PAGE
Quantity 100 μg
Source E. coli
Storage Requirements -80°C for 12 months
Cross Reactivity Human
Recombinant Recombinant
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.