Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ NAP1 Active Protein

Catalog No. 14837 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
300 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-837 300 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-837 Supplier MilliporeSigma™ Supplier No. 14837
Add to Cart
Add to Cart

Specifications

Characterization 90% by SDS-PAGE
Formulation 10mM K-HEPES pH 7.6, 10mM KCl, 50mM NaCl, 0.1mM EDTA, 10%(v/v) glycerol, 0.01% NP-40, 0.2mM PMSF, 1mM DTT, Frozen solution.
Gene ID (Entrez) 37798
Name NAP1 Active Protein
Purification Method Ni2+/NTA-agarose
Quantity 300 μg
Source E. coli
Storage Requirements -80°C for 6 months
Conjugate N-terminal His-Tag
Cross Reactivity Drosophila
Recombinant Recombinant
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.