Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate p300, HAT Domain Recombinant Enzyme

Catalog No. 14418 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-418 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-418 Supplier MilliporeSigma™ Supplier No. 14418
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant GST-fusion protein corresponding to amino acids 1066-1707 of human p300.

EP300 encodes the adenovirus E1A-associated cellular p300 transcriptional co-activator protein. p300 is related by sequence to CBP (CREB-binding protein [CREB: cyclic-AMP responsive element binding protein]), and like CBP can stimulate transcription through activation of CREB. This EP300 activity is specifically inhibited by the adenovirus oncoprotein E1A. EP300 has also been identified as a co-activator of HIF1A (hypoxia-inducible factor 1 alpha), and thus plays a role in the stimulation of hypoxia-induced genes such as VEGF.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Histone Acetyltransferase Assay (HAT)
Content And Storage -20°C for up to 6 months from the date of receipt
Source Expressed in E. coli
Quantity 50 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.