Learn More
Description
Includes
Components of Kit
- 100X GDP, 100mM
- Rac/cdc42 Assay Reagent (PAK1 PBD, agarose)
- Anti-Rac2
- Mg2+ Lysis/Wash Buffer, 5X
- 100X GTP?S, 10mM
Specifications
Specifications
| For Use With (Application) | Affinity Binding Assay |
| Includes | 100X GDP 100mM, Rac/cdc42 Assay Reagent (PAK1 PBD, agarose), Anti-Rac2, Mg2+ Lysis/Wash Buffer 5X, 100X GTPγS 10mM |
| Quantity | 30 Assays |
| Type | Activation Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.