Learn More
MilliporeSigma™ Upstate RalB Activation Assay Kit (Non-radioactive)
Shop All MilliporeSigma ProductsDescription
- Suitable for G-protein affinity-binding assays.
Specifications
Specifications
| Content And Storage | -20°C for 12 months |
| Description | RalB Activation Assay Kit (Non-radioactive) |
| For Use With (Application) | Non-radioactive Ral-B Activation Assay Kit can be used to precipitate Ral-B-GTP from cell lysates and detection by a Ral-B specific monoclonal antibody. |
| Includes | 100X GDP (100mM), anti-RalB, clone 25, Ral Assay Reagent, Mg2+ Lysis/Wash Buffer (5X), 100X GTPyS (10mM) |
| Quantity | 20 Assays |
| Type | Activation Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.