Specifications
| Description | Class III HDAC Assay Kit |
| For Use With (Application) | Enzyme Assay |
| Includes | Developer Reagent, Black 96 well plate and covers, SIRTaintyTM Assay Buffer, Acetylated Peptide Substrate, β-NAD, Nicotinamide (NAM), Recombinant Sirt1 Enzyme, Recombinant Nicotinamidase Enzyme, Suramin (Sirt Inhibitor) |
| Quantity | 96 Assays |