Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ VEGF Human Recombinant Protein

Catalog No. 01185MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-185MI 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-185MI Supplier MilliporeSigma™ Supplier No. 01185
Add to Cart
Add to Cart

Specifications

For Use With (Application) Cell Culture
Name VEGF Human Recombinant Protein
Quantity 10 μg
Source Recombinant human DNA expressed in baculovirus infected Sf9 cells
Cross Reactivity Human
Recombinant Recombinant
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.