Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ VEGFR 2D1-7, Human, Recombinant Protein, S. frugiperda

Catalog No. 67649050UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-649-050UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-649-050UG Supplier MilliporeSigma™ Supplier No. 67649050UG
Add to Cart
Add to Cart
Recombinant, human VEGFR-2 consisting of all 7 immunoglobulin-like extracellular domains, but lacking the single transmembrane region and the intracellular split tyrosine kinase domain expressed in S. frugiperda insect cells. Binding of ligand to the full-length VEGFR-2 results in a high signaling activity and mitogenic activity in endothelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 100mM NaCl, 25mM MES, pH 5.5.
Preparation Method Aqueous buffers (≥100μg/mL) or H2O (≥100μg/mL)
Purification Method SDS-PAGE
Quantity 50 μg
Source S. frugiperda
Immunogen VEGFR2D1-7
Storage Requirements Following reconstitution, aliquot and freeze (−70°C). Stock solutions are stable for up to 6 months at −70°C. Avoid freeze/thaw cycles of solutions.
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.