Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ von Willebrand Factor, Human Plasma, Protein

Catalog No. 681300100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-130-0100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 68-130-0100UG Supplier MilliporeSigma™ Supplier No. 681300100UG
Add to Cart
Add to Cart
Native, von Willebrand factor from human plasma. Glycoprotein complex found in plasma, platelets, and subendothelium. An adhesive protein that mediates the initiation and progression of throm bus formation at the site of injury.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 100mM glycine, 100mM NaCl, 25mM sodium citrate, pH 6.8.
Preparation Method H2O
Purification Method SDS-PAGE
Quantity 100 μg
Immunogen von Willebrand Factor
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 1 year at −20°C.
Regulatory Status RUO
Species Human
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.