Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Xanthine Oxidase, Buttermilk, Protein

Catalog No. 68215110U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 U
50 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-215-110U 10 U
68-215-150U 50 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 68-215-110U Supplier MilliporeSigma™ Supplier No. 68215110U
Add to Cart
Add to Cart
Native xanthine oxidase from buttermilk. Complex metalloprotein that catalyzes oxidative hydroxylation of a variety of aromatic heterocycles and simple aldehydes. Catalyzes the oxidation of xanthine to uric acid.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation Crystalline suspension in 60% ammonium sulfate.
Quantity 10 U
Immunogen Xanthine Oxidase
Regulatory Status RUO
Species Bovine (Buttermilk)
Recombinant Native
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.