Learn More
Apexbio Technology LLC EMD638683, 10mg Cas No: 1181770-72-8 MFCD: MFCD25541743
SDP
Supplier:  Apexbio Technology LLC A338910
Encompass_Preferred
EMD638683 is a highly selective inhibitor of the serum and glucocorticoid inducible kinase (SGK) with IC50 value of 3 M [1].SGK is a group of serine/threonine kinases that consists of SGK1, SGK2 and SGK3. There are other sizes available. Please inqury us for quote.
Catalog No. NC1569479