Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMDT oxalate, Tocris Bioscience™

Catalog No. 420950
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-095-0 50 mg
42-091-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-095-0 Supplier R&D Systems Supplier No. 4209/50
This item is not returnable. View return policy

Selective 5-HT6 agonist

Selective 5-HT6 agonist (with a Ki value of 16 nM at human 5-HT6 receptors). Exhibits potency comparable to serotonin for the activation of adenylate cyclase. Displays antidepressant-like effects.

Chemical Identifiers

CAS 263744-72-5
Molecular Formula C15H22N2O
Molecular Weight (g/mol) 246.354
InChI Key ZEYRDXUWJDGTLD-UHFFFAOYSA-N
Synonym emdt, edmt, 2-2-ethyl-5-methoxy-1h-indol-3-yl-n,n-dimethylethanamine, 2-ethyl-5-methoxy-n,n-dimethyltryptamine, d02ool, d0ck9k, gtpl12, n,n-dimethyl-2-ethyl-5-methoxy-1h-indole-3-ethanamine, 2-2-ethyl-5-methoxy-1h-indol-3-yl ethyl dimethylamine, 6
PubChem CID 6918513
IUPAC Name 2-(2-ethyl-5-methoxy-1H-indol-3-yl)-N,N-dimethylethanamine
SMILES CCC1=C(C2=C(N1)C=CC(=C2)OC)CCN(C)C

Specifications

Quantity 50 mg
Formula Weight 336.38
Percent Purity >98%
Chemical Name or Material EMDT oxalate
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.