Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Emerald CX™ Nylon/Stainless Steel Knit Cut-Resistant Gloves

Catalog No. 17401210
Change view
Click to view available options
Size:
5
6
7
8
9
10
11
12
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-401-210 8
17-401-207 5
17-401-208 6
17-401-209 7
17-401-211 9
17-401-212 10
17-401-213 11
17-401-214 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 17-401-210 Supplier Superior Glove™ Supplier No. S13KBPU8
Add to Cart
Add to Cart
This item is not returnable. View return policy

Nylon and fine-gauge steel cut-resistant gloves with polyurethane palm coating provides a secure grip, dexterity and touch sensitivity.

  • Combination of nylon and fine-gauge steel
  • Lint-free nylon is lightweight and tough for better dexterity
  • Comfortable with touch sensitivity
  • Polyurethane palm coating provides secure grip and tactile feel
  • Ideal for glass handling, non-marring gloves will not leave residue, smudges or fingerprints
  • Extra-long knit wrists provide extra protection
  • Touchscreen compatible

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A2
Material Nylon, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size 8
For Use With (Application) HVAC, automotive, sharp Steel, Glass, Metal fabrication, light Metal stamping
Glove Cuff Style Knit
Model S13KBPU/8
Puncture Rating ANSI/ISEA 105-16 Level 2
Series Emerald CX™
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.