Description
*NOTE: Emerge frame must be presented to your optometrist in order to be fitted with prescription lenses
Specifications
Specifications
| Certifications/Compliance | ANSI Z87.1,CSA Z94.3-07 |
| Description | Will accommodate an Rx lens to the maximum of +3 to -3 diopters |
| Lens Tint | IR/PC 3.0 |
| Frame Color | Black |
| Magnification Power | None |
| Weight (Metric) | 30 g |
| Lens Material | Polycarbonate |
| Frame Material | G300 Nylon |