Learn More
Poltex Emergency Airway Procedure Storage Box

Description
- Emergency crash box holds resuscitation bag and tubing, manual ventilators, etc.
- Magnetic closure and stainless steel hinges
- Front latch accepts either a locking safety seal, or a padlock
- Made from clear PETG
- Choose from two sizes
- Available as magnet mount (powerful magnets included), wall mount (screws/anchors included), and tape mount (VHB tape included)
Specifications
Specifications
| Product Type | Emergency Airway Procedure Storage Box |
| Color | Clear |
| Material | PETG |
| Dimensions (L x W x H) | 8 x 16 x 20 in. |
| For Use With (Equipment) | Resuscitation Bag and Tubing |
| Style | Magnetic Mount |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.