Learn More
Description
- Contains Narcan to rapidly reverse life-threatening opioid effects.
- Includes CPR mask for safe assisted breathing during emergencies.
- 2 x 2 gauze pad for quick cleanup or nasal application.
- Antiseptic wipe to disinfect before or after administering aid.
- Chemical-resistant nitrile gloves to ensure hygienic handling and protection.
Specifications
Specifications
| Product Type | Overdose Kit |
| Color | White |
| Material | Plastic |
| Disposable | Single-use |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.