Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Emetine Dihydrochloride Hydrate, TCI America™
SDP

Catalog No. E00071G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E00071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E00071G Supplier TCI America Supplier No. E00071G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 316-42-7
Molecular Formula C29H42Cl2N2O4
Molecular Weight (g/mol) 553.565
MDL Number MFCD00135589
InChI Key JROGBPMEKVAPEH-GXGBFOEMSA-N
Synonym emetine dihydrochloride, emetine, dihydrochloride, emetine hydrochloride, purum, emetine hcl, emetine.2hcl, amebicide, emetine dihydrochloride hydrate, l-emetine dihydrochloride, unii-u52og12p96
PubChem CID 3068143
IUPAC Name (2S,3R,11bS)-2-[[(1R)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-1-yl]methyl]-3-ethyl-9,10-dimethoxy-2,3,4,6,7,11b-hexahydro-1H-benzo[a]quinolizine;dihydrochloride
SMILES CCC1CN2CCC3=CC(=C(C=C3C2CC1CC4C5=CC(=C(C=C5CCN4)OC)OC)OC)OC.Cl.Cl

Specifications

pH 5.6 (1g/50mL H2O soln.)
UN Number 1544
Quantity 1 g
Formula Weight 553.57
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Emetine Dihydrochloride Hydrate
RTECSNumber JY5250000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.