Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ EMMPRIN/CD147 Antibody Pair [HRP]
SDP

Catalog No. NB010015 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010015 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010015 Supplier Novus Biologicals™ Supplier No. NBP27948815PLATES
Add to Cart
Add to Cart

Monoclonal Antibody Pair

specifically detects in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 6.25 pg/mL
Assay Sensitivity 6.25-400 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human EMMPRIN/CD147.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias 5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor
Gene ID (Entrez) 682
Gene Symbol BSG
Host Species Rabbit/Mouse
Kit Contents
  • Standard: Each vial contains 25 ng of recombinant EMMPRIN/CD147.
  • Capture Antibody: 0.5 mg/mL of mouse anti-EMMPRIN/CD147 monoclonal antibody,
  • Detection Antibody: 0.5 mg/mL rabbit anti-EMMPRIN/CD147 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cancer, Signal Transduction, Vision
    Storage Requirements Storage is content dependent.
    Target EMMPRIN/CD147
    Technique 1. Capture Antibody: Dilute to a working concentration of 4.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.125 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 400 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.