Learn More
Selleck Chemical LLC EMPA-E0107-5MG
SDP
Supplier:  Selleck Chemical LLC E01075MG
Encompass_Preferred
EMPA is a selective OX2 receptor antagonist and binds to human and rat OX2-HEK293 membranes with Kd values of 1 1 and 1 4 nM respectively
Catalog No. 50-234-0770
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.