Learn More
Cayman Chemical Empagliflozin, 864070-44-0, MFCD22566222, 50mg
SDP
Supplier:  Cayman Chemical 1737550
Encompass_Preferred
Molecular Formula: C23H27ClO7, Purity: ≥98%, Formula Weight: 450.9, Synonyms: BI 10773, (1S)-1,5-anhydro-1-C-[4-chloro-3-[[4-[[(3S)-tetrahydro-3-furanyl]oxy]phenyl]methyl]phenyl]-D-glucitol; Inhibiting renal glucose reabsorption through the sodium-glucose cotransporter (SGLT) offers an insulin-independent alternative to controlling blood glucose concentrations in patients with type 2 diabetes.
Catalog No. NC1613643
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.