Learn More
Enzo Life Sciences EMPO (50mg)
SDP
Supplier:  Enzo Life Sciences ALX430098M050
Encompass_Preferred
Beside DEPMPO (ALX-430-093), most efficient spin trap for the in vitro and in vivo detection of O-, N-, S-, and C-centered free radicals. Has a longer life-time than DMPO (ALX-430-090). Alternative name: 2-Ethoxycarbonyl-2-methyl-3,4-dihydro-2H-pyrrole-1-oxide. Purity: ≥95% (HPLC). Appearance: Colorless to light yellow oil. Solubility: Soluble in water, DMSO, ethanol or dichloromethane. Long Term Storage: -80°C.
Catalog No. 50-200-9202
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.