Description
CDS™ Empore™ StageTips with C18/SCX/C18 layers, C18 and cation exchange (SCX) solid phase extraction micropipette tips.
- Combines good retention capacity with high recoveries
- Recommended for desalting of large peptides and proteins and peptide fractionation at acidic, neutral, and basic pH conditions
Specifications
Specifications
| Particle Size | 12 μm |
| Product Line | Empore |
| Sorbent | DVB (Polystyrene-divinylbenzene) |
| Volume (Metric) | 200 μL |
| Quantity | 96 Pk. |