Learn More
CDS Analytical™ Empore™ C18/SCX StageTips
SDP
Catalog No. 13110068 Shop All CDS Analytical Products
Change view
Click to view available options
Quantity:
96 Pk.
960 Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-110-068 960 Cs.
13-110-067 96 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 13-110-068 Supplier CDS Analytical™ Supplier No. 70201910076
Add to Cart
Add to Cart
This item is not returnable. View return policy
C18 bonded silica and cation exchange (SCX) StageTips are SPE micropipette tips for the desalting of large peptides and proteins, as well as peptide fractionation.
CDS™ Empore™ StageTips C18 bonded silica and cation exchange (SCX) solid phase extraction micropipette tips combine good retention capacity with high recoveries.
  • Recommended for desalting of large peptides and proteins and peptide fractionation at acidic, neutral, and basic pH conditions
Specifications
Particle Size 12 μm
Product Line Empore
Sorbent DVB (Polystyrene-divinylbenzene)
Volume (Metric) 200 μL
Quantity 960 Cs.