Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Empore™ Extraction Disk Cartridge, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022327 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric) Inner:
4 mm
7 mm
10 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
11-102-2327 10 mm
11-102-2325 4 mm
11-102-2326 7 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-102-2327 Supplier MilliporeSigma Supelco Supplier No. 66873U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recommended for the extraction of moderately-polar to non-polar analytes from aqueous samples (e.g., biological plasma).

  • Empore membrane SPE technology comprises of SPE particles tightly enmeshed within a network of inert PTFE fibrils (90% SPE particles : 10% PTFE, by weight).
  • The SPE-membrane fabrication process results in a highly dense and uniform extraction medium that offers distinct advantages over traditional sorbent/packed-bed SPE products.
  • Empore SPE technology provides a denser, more uniform extraction bed than traditional packed bed products allowing for smaller bed weights, shorter analyte to pore diffusion paths, and more efficient extractions.
  • Layed above the SPE membrane is a polypropylene pre-filter to prevent particulates from reaching the underlying membrane. The dense particle packing and uniform distribution within the Empore membrane offers outstanding extraction efficiency and reproducibility.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 30/Pk.
Particle Size 50 μm
Pore Size 60 Å
Diameter (Metric) Inner 10 mm
Stationary Phase C18-SD
Material Polypropylene filter (graded density pre-filter)
Endcapped Endcapped

Empore is a trademark of CDS Analytical

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.