Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDS Empore™ Filter Aid
SDP

Catalog No. 13110009 Shop All CDS Analytical Products
Change view
Click to view available options
Quantity:
1.5 kg/Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-110-009 1.5 kg/Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-110-009 Supplier CDS Supplier No. 98050300767
Add to Cart
Add to Cart

Formerly 3M™ Empore™ FAID400 is a filtration aid when extracting samples with a high particulate content using Empore SPE disks

  • 1.5 kg/bottle with dispensing scoop
  • Use: Approximately 40 grams will cover a 90 mm disk
    1 cm deep
  • Approximately 15 grams will cover a 47 mm
    disk 1 cm deep
  • Material: Glass beads; average diameter 40 microns
  • Disposal: If no hazardous, as lab glassware; if hazardous, follow regulations

Specifications

Base Material Glass Bead
For Use With (Application) As a Filtration Aid When Extracting Samples with a High Particulate Content
Particle Size 40 μm
Type Filter Aid
Quantity 1.5 kg/Each
Length (Metric) 155 mm
Diameter (Metric) Outer 75 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.