Learn More
CDS Analytical™ Empore™ SDB-XC StageTips
SDP
Catalog No. 13110059 Shop All CDS Analytical Products
Change view
Click to view available options
Quantity:
96 Pk.
960 Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-110-059 96 Pk.
13-110-060 960 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 13-110-059 Supplier CDS Analytical™ Supplier No. 70201910037EA
Add to Cart
Add to Cart
This item is not returnable. View return policy
SDB-XC (styrenedivinylbenzene) polymer sorbent StageTips are SPE micropipette tips for the desalting of large peptides and proteins, as well as peptide fractionation.
The CDS™ Empore™ StageTips SDB-XC (styrenedivinyl benzene) polymer sorbent solid phase extraction micropipette tips have two layers for increased capacity.
  • Combines good retention capacity with high recoveries
  • Recommended for desalting of large peptides and proteins and peptide fractionation at acidic, neutral, and basic pH conditions
Specifications
Particle Size 12 μm
Product Line Empore
Sorbent DVB (Polystyrene-divinylbenzene)
Volume (Metric) 200 μL
Quantity 96 Pk.