Learn More
JDM SCIENTIFIC LLC Empty 2D bC SBS racks
Supplier:  JDM SCIENTIFIC LLC 202500
Encompass_Preferred
Empty large format 81 rack for 1 8ml 1 2ml 2ml 2D barcoded storage tubes AlteClean 3 racks per resealable bag 4 resealable bags per case
Catalog No. 50-000-14257
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.