Learn More
Sigma Aldrich Empty Cartridges 3 Ml Opaque Black 50/box
Catalog No. 9175161EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
91-751-61EA 1 Ea.
1 options
Catalog No. 91-751-61EA Supplier Sigma Aldrich Supplier No. 9175161EA
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Empty cartridges for 3D Bioprinting application. The precision fit between the barrel and piston ensures consistent fluid deposition. The wiper piston improves fluid control and prevents fluid from dripping. Pistons, end and tip caps are sold separately.Opaque black cartridges for complete light blockage
Specifications
Quantity 1 Ea.
Synonym 3d bioprinting consumables; Syringe barrels