Learn More
Fisherbrand™ Empty Click Lock Container
SDP
Catalog No. 23101010 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
20 mL
40 mL
90 mL
120 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
23-101-010 40 mL
23-101-009 20 mL
23-101-012 90 mL
23-101-013 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-101-010
Add to Cart
Add to Cart
This item is not returnable. View return policy
Empty Click Lock Container designed for safe, leak-resistant transport and storage of fluids.
  • Designed for safe, leak-resistant transport and storage of fluids
  • Screw-top containers have knurled cap and base for easy handling
  • Containers are graduated and made of durable clarified polypropylene
Specifications
Product Type Specimen Container Vial
Closure Included Yes
Graduated Yes
Material Polypropylene
Shape Cylindrical
Sterility Non-sterile
Autoclavable Not Autoclavable
Capacity (Metric) 40 mL
Color Clear
No. per Case 800