Learn More
MilliporeSigma™ Supelco™ Empty Reversible SPE Tube, Non-fluorous Polypropylene, PE frit included
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Column Type | SPE |
| Description | Empty, Use PE frits for 3 mL PP SPE tube size (57180-U) |
| For Use With (Application) | Food and Beverages |
| Type | Solid Phase Extraction Column |
| Volume (Metric) | 1 mL |
| Quantity | 50/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.