Learn More
MilliporeSigma™ Supelco™ Empty SPE Tube (No Frits)
Empty SPE Tubes allow you to pack your own SPE cartridges
Supplier: MilliporeSigma™ Supelco™ 57179
Description
When used in conjunction with SPE Tube Adapters, empty SPE tubes can be stacked on top of a standard SPE cartridge during processing. The empty SPE cartridge acts as a reseroir to accommodate larger sampling volumes.
Specifications
SPE | |
Food and Beverages | |
12 mL | |
20/Pk. |
Empty | |
Solid Phase Extraction Column | |
Polypropylene |