Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Empty Stainless Steel GC Column

Catalog No. 111011282 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
2.4 m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-101-1282 2.4 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-1282 Supplier MilliporeSigma™ Supelco™ Supplier No. 13097U

missing translation for 'validMainframeMsg'

For less demanding applications where the inertness of glass is not required, choose more economical stainless steel. Column can be carefully bent to fit most instruments.

Empty column may be used for packing adsorbents for temperature-programmed desorption (TPD) instrumentation.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Gas Chromatography (GC)
Length (Metric) 2.4 m
Material Stainless Steel Column
Packing Material None
Type Empty GC
Diameter (Metric) Inner 2.1 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.