Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EMR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102076
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102076 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102076 Supplier Invitrogen™ Supplier No. PA5102076
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total EMR4.

FIRE is a 689 amino acid protein that maps to human gene EMR4. FIRE is also known as EGF-like module-containing mucin-like hormone receptor-like 4 and is a member of the the G protein-coupled receptor 2 family, LN-TM7 subfamily. The epidermal growth factor-transmembrane seven (EGF-TM7) family are proteins that express EGF-like domains at their extracellular N-terminus coupled to a classical seven-transmembrane (TM7) cassette. FIRE is believed to mediate the cellular interaction between myeloid cells and beta cells. FIRE is a membrane bound, multi-pass protein that is proteolytically cleaved into two subunits, an extracellular alpha subunit and a seven-transmembrane subunit. FIRE contains one GPS (G protein-coupled receptor proteolytic cleavage site) domain and two EGF (epidermal growth factor)-like domains, the second of which mediates the interaction with the putative ligand.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMR4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADGRE4P
Gene Accession No. Q86SQ3
Gene Alias ADGRE4P; adhesion G protein-coupled receptor E4, pseudogene; EMR4; EMR4P; FIRE; GPR127; PGR16
Gene Symbols ADGRE4P
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADGRE4P(Accession Q86SQ3), corresponding to amino acid residues K129-L179.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 326342
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.