Learn More
Selleck Chemical LLC Emricasan 5mg 254750-02-2 IDN-6556, PF 03491390, PF-03491390
SDP
Supplier:  Selleck Chemical LLC S77755MG
Encompass_Preferred
Emricasan (IDN-6556, PF 03491390, PF-03491390) is a potent irreversible pan-caspase inhibitor. Emricasan is an inhibitor of Zika virus infection. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5234
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.