Learn More
Electron Microscopy Sciences EMS-Mount 1000 ML
SDP
Supplier:  Electron Microscopy Sciences 1798508
Encompass_Preferred
EMS-Mount is a non-permanent, low viscosity an aqueous mounting medium designed for mounting of tissue sections and cell smears with peroxidase and alkaline phosphatase chromogens that cannot be dehydrated with organic solvents. This mounting medium preserves Fast red, Aminoethylycarbazole (AEC), BCIP/NBT, BCIP/INT chromogens and is also compatible with counterstain like Hematoxylin and Nuclear fast red (NFR). It is also suitable for chromogens like DAB and DAB with nickel and cobalt. (It is not compatible with H and E staining).
Application: Mounting of chromogens that cannot be dehydrated with organic solvents. Ready to use. Storage at 2-8 degree C is recommended.
Catalog No. 50-343-72
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.