Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Emulsifying Wax, NF, Spectrum™ Chemical
SDP

Catalog No. 18602407 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
12 x 500 g
25 kg
Packaging:
Fiber Drum
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-407 12 x 500 g Poly Bottle
18-602-406 500 g Poly Bottle
18-602-408 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-602-407 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. W1026500GMCS12
Add to Cart
Add to Cart

W1026

Spectrum™ Chemical Emulsifying Wax, NF is used as a thickening agent excipent. All Spectrum Chemical NF products are packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

CAS Max % 100%
Melting Range 50-54°C
Packaging Poly Bottle
Quantity 12 x 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.