Learn More
Medchemexpress LLC HY-134761 5mg , EN4 CAS:1197824-15-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1347615MG
Encompass_Preferred
HY-134761 5mg EN4 CAS: 1197824-15-9 EN4 is a covalent ligand that targets cysteine 171 (C171) of MYC. EN4 is selective for c-MYC over N-MYC and L-MYC. EN4 inhibits MYC transcriptional activity, downregulates MYC targets, and impairs tumorigenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0755