Learn More
Medchemexpress LLC Enalaprilat-d5 | 349554-00-3 | 99.5% | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0231AS10MG
Encompass_Preferred
Enalaprilat-d5 is a deuterium-labeled analogue of enalaprilat used as a reference standard in biochemical and pharmacokinetic research. It acts as an angiotensin-converting enzyme (ACE) inhibitor and is provided as a high-purity analytical standard for assay development and quantitation.
  • Deuterium-labeled internal standard for analytical assays.
  • High isotopic enrichment and 99.5% purity for reliable quantitation.
  • Suitable for LC-MS/MS method development and validation.
  • Available in small pack sizes (for example, 10 mg) for bench-scale work.
  • Supplied as a dry powder for convenient dissolution and storage.
Catalog No. 50-002-97697
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.