Learn More
Medchemexpress LLC HY-109057 10mg , Enarodustat CAS:1262132-81-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10905710MG
Encompass_Preferred
HY-109057 10mg Enarodustat CAS: 1262132-81-9 Enarodustat is a potent and orally active hypoxia-inducible factor prolyl hydroxylase inhibitor, with an EC50 of 0.22 μM. Enarodustat has the potential for renal anemia treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0500
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.