Learn More
Medchemexpress LLC HY-18690 10mg Medchemexpress, Enasidenib CAS:1446502-11-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1869010MG
Encompass_Preferred
Medchemexpress, HY-18690 10mg Enasidenib CAS:1446502-11-9 Enasidenib is an oral, potent, reversible, selective inhibitor of the IDH2 mutant enzymes, with IC50s of 100 and 400 nM against IDH2R140Q and IDH2R172K, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9265
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.