Learn More
Ansell ENCORE™ Latex Micro Surgical Gloves
GREENER_CHOICE
Catalog No. 19501135 Shop All Ansell Products
Change view
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-501-135 5.5
19-501-136 6
19-501-137 6.5
19-501-138 7
19-501-139 7.5
19-501-140 8
19-501-141 8.5
19-501-142 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-501-135 Supplier Ansell Supplier No. 5787000
Add to Cart
Add to Cart
Thin surgical gloves feature a soft latex formulation and a smooth micro-textured finish for enhanced dexterity and tactile sensitivity.
  • Thinner than standard latex powder free surgical gloves
  • Brown color reduces glare of surgical lights
  • Suitable to be used as an under glove
  • DERMASHIELD™ Technology facilitates dry or damp donning and intra-operative changes
  • Recommended for all surgeries especially delicate procedures in ophthalmology, micro and cardiovascular
  • Enhanced tactile sensitivity and dexterity
  • Prevention of eye fatigue
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM D3577, ASTM D7160, ASTM D6978-05
Certifications/Compliance (Europe) EN 16523-1, EN 374:2003, EN 420:2003 + A1:2009, EN 455 1-4, EN ISO 374-1:2016, EN ISO 374-5:2016, ISO 10282
AQL 0.65
Color Brown
Exterior Finish No Texture
Laboratory/Cleanroom Use No
Material Latex
Size 5.5
Sterility Sterile
Manufacturing/QMS Audit Standards EN 556, ISO 11137-Part 1, ISO 13485, ISO 14001, ISO 9001
Model 5787000
Series ENCORE™ Latex Micro
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.