Learn More
Ansell ENCORE™ Perry Style 42™ PF Surgical Gloves
Catalog No. 19501156 Shop All Ansell Products
Change view
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-501-156 8
19-501-151 5.5
19-501-152 6
19-501-153 6.5
19-501-154 7
19-501-155 7.5
19-501-157 8.5
19-501-158 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-501-156 Supplier Ansell Supplier No. 5711105PF
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Latex, powder free surgical gloves feature a smooth finish and optimized thickness. Ideal for brief, low-risk sterile procedures inside or outside of the operating room.
  • Natural rubber latex
  • Made with proprietary anatomic formers for comfort and to help reduce hand fatigue
  • Polymer coated for fast and easy donning
  • Optimized glove thickness for the best balance of protection and tactile sensitivity
  • Recommended for:
    • Brief, low-risk examination procedures
    • Sterile procedures in the O.R.
    • Use as an outer glove when double gloving
    • Sterile procedures outside of the O.R.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM D3577
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
AQL 0.65
Color Natural
Exterior Finish No Texture
Laboratory/Cleanroom Use No
Material Latex
Size 8
Sterility Sterile
Manufacturing/QMS Audit Standards ISO 13485, ISO 9001
Model 5711105PF
Series ENCORE™ Perry Style 42™ PF
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.