Learn More
Thermo Scientific™ Click-On™ Inline Gas Filter Connectors
Catalog No. 60180809
Change view
Click to view available options
Type:
End Fitting
Replacement Filter O-Ring
Steel End Fitting
Wall Mounting Clamp
Material:
Brass
No Information Available
Steel
Diameter (English) Outer:
0.125 in.
0.25 in.
N/A
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Material Diameter (English) Outer
60180809 End Fitting Brass 0.125 in.
60180810 Steel End Fitting Steel 0.125 in.
60180811 End Fitting Brass 0.25 in.
60180812 Steel End Fitting Steel 0.25 in.
60180834 Wall Mounting Clamp No Information Available N/A
60180833 Replacement Filter O-Ring No Information Available N/A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60180809 Supplier Thermo Scientific™ Supplier No. 60180809
Add to Cart
Add to Cart
Use these quality end fittings o-rings, and other connectors for your Thermo Scientific™ Click-On™ Inline Filters.
Use these quality end fittings o-rings, and other connectors for your Thermo Scientific™ Click-On™ Inline Filters. The end fittings remain installed during filter replacement, thereby reducing the cost of clean gas.
  • Incorporates needle valves that close lines and prevent contamination
  • End fittings measure 0.125in. (0.3cm) or 0.25in. (0.6cm)
  • Reusable
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Click-On Inline Filter
Diameter (English) Outer 0.125 in.
Material Brass
Type End Fitting
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.