Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

End-Repair dA-tailing Mix, 24 reactions, G-Biosciences
SCI_ED

Catalog No. S001163
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
S001163 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001163 Supplier G-Biosciences Supplier No. 7862279
Add to Cart
Add to Cart

The End-Repair dA-tailing Mix converts damaged or incompatible 5'/3′ ends to 3' A tailed and 5′-phosphorylated DNA.

  • This Mix uses T4 DNA Polymerase's 5'→3' polymerase and 3'→5' exonuclease actions to blunt the fragment ends before Taq DNA polymerase adds an A Tail at the 3' end and T4 Polynucleotide Kinase ensures 5' phosphorylation.

Specifications

Packaging Vial
Enzyme Polymerases and Kinase
Sensitive to Heat Inactivation Yes
Product Type End-Repair dA-tailing Mix
Research Category Next-generation Sequencing, DNA Repair, Traditional Cloning
Product Line Thermo Scientific
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.