Specifications
| Lens Coating | Scratch-resistant |
| Certifications/Compliance | ANSI Z87.1,CSA Z94.3-07 |
| Lens Tint | Clear |
| Frame Color | Black, Green |
| Weight (Metric) | 23.5 g |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate, TPE |