Description
Certifications
UL listed, CSA, CE marked
Compliance
Meets all relevant OSHA and ANSI Z9.5 standards
Specifications
Specifications
| Product Type | Ductless Fume Hood |
| Dimensions (D x W x H) | 26 x 35 x 35 in. (66 x 89 x 89 cm) |
| For Use With (Equipment) | Ductless enclosures |
| Includes | AirSafe NXT color touch screen; Fully assembled; Filters installed |