Learn More
BioVendor, LLC EndoGrade Endotoxin-free Ovalbumin, 100 mg.
SDP
Supplier:  BioVendor, LLC LET0028
Encompass
EndoGrade Endotoxin-free Ovalbumin, 100 mg. Endotoxin level < 0.1 EU/mg. Lyophilized. Contains no salts. Purity > 98%. Molecular weight: ~ 45 kDa. Source: Chicken Egg White. Ovalbumin is an extensively used allergen model in immunology and allergology studies. Commercially available ovalbumin is often contaminated with endotoxin, which leads to artefacts and inaccurate results. Endotoxin-free EndoGrade Ovalbumin is of superior quality with a guaranteed endotoxin level (LPS) below 0.1 EU/mg. EndoGrade Ovalbumin is manufactured in Germany by LIONEX GmbH and has been referenced in over 200 scientific publications.
Catalog No. 50-206-9086
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.