Learn More
World Precision Instrument Cell Culture Cup Chambers for TEER Measurement-ENDOHM-6G
SDP
Supplier:  World Precision Instrument ENDOHM6G
Encompass
These Cell Culture Cup Chambers used in TEER measurement for removable culture cup systems using EVOM2 meters for endothelial and epithelial cell cultures.
NOTE: These EndOhms are for use with older EVOM™ meters. For EndOhms used with the new EVOM™ Manual (or EVOM3), see EVOM™ Electrodes for TEER.
EndOhm chambers provide reproducible resistance measurements of endothelial and epithelial monolayers in culture cups. Transfer cups from their culture wells to the EndOhm chamber for measurement rather than using hand-held electrodes. The chamber and the cap each contain a pair of concentric electrodes: a voltage-sensing silver/silver chloride pellet in the center plus an annular current electrode.
Catalog No. NC0210128
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.